1-(2-Chloro-6-hydroxyphenyl)ethanone 97%

Supplier: Ambeed
Warning

A328859-100MG A328859-10G A328859-1G A328859-250MG A328859-25G A328859-5G
76788-150EA 7.43 USD
76788-150 76788-152 76788-154 76788-156 76788-158 76788-160
1-(2-Chloro-6-hydroxyphenyl)ethanone 97%
1-(2-Chloro-6-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00218550
CAS Number: 55736-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR