TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) 99%

Supplier: Ambeed
Warning

Synonyms: TAPSO, 2-Hydroxy-3-{[2-hydroxy-1,1-bis(hydroxymethyl)-ethyl]amino}-1-propanesulphonic acid, 3-[N-Tris(hydroxymethyl)methylamino]-2-hydroxypropanesulphonic acid

A327675-100G A327675-25G A327675-5G
76788-144EA 44 USD
76788-144 76788-146 76788-148
TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) 99%
TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)

Precaución: for Research Use Only

Formula: C₇H₁₇NO₇S
MW: 259.28 g/mol
MDL Number: MFCD00038352
CAS Number: 68399-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR