4-(Pyrrolidin-1-yl)-3-(trifluoromethyl)aniline dihydrochloride 95+%

Supplier: Ambeed
Warning

A327219-1G A327219-250MG A327219-5G
76788-138EA 73.21 USD
76788-138 76788-140 76788-142
4-(Pyrrolidin-1-yl)-3-(trifluoromethyl)aniline dihydrochloride 95+%
4-(Pyrrolidin-1-yl)-3-(trifluoromethyl)aniline dihydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅Cl₂F₃N₂
MW: 303.1500 g/mol
MDL Number: MFCD04117872
CAS Number: 1158215-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR