Tetramethylammonium iodide 99%

Supplier: Ambeed
Warning

A325725-100G A325725-25G A325725-500G A325725-5G
76788-120EA 35 USD
76788-120 76788-122 76788-124 76788-126
Tetramethylammonium iodide 99%
Tetramethylammonium iodide

Precaución: for Research Use Only

Formula: C₄H₁₂IN
MDL Number: MFCD00011629
CAS Number: 75-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR