tert-Butyl-2,4-dioxopiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2,4-dioxopiperidine-1-carboxylate, Piperidine-2,4-dione, N-BOC protected

A325155-100G A325155-10G A325155-1G A325155-25G A325155-5G A325155-500G
76788-106EA 140 USD
76788-106 76788-108 76788-110 76788-112 76788-114 77872-466
tert-Butyl-2,4-dioxopiperidine-1-carboxylate 97%
tert-Butyl-2,4-dioxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₄
MDL Number: MFCD10566071
CAS Number: 845267-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR