Tetraethylene glycol monododecyl ether 98%

Supplier: Ambeed
Danger

Synonyms: 3,6,9,12-Tetraoxatetracosan-1-ol, PEG 4 lauryl ether

A324520-100MG A324520-1G A324520-250MG
76788-092EA 17 USD
76788-092 76788-094 77872-460
Tetraethylene glycol monododecyl ether 98%
Tetraethylene glycol monododecyl ether

Precaución: for Research Use Only

Formula: C₂₀H₄₂O₅
MDL Number: MFCD00043063
CAS Number: 5274-68-0
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR