4-Phenylethynylbenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Phenylethynyl)benzaldehyde

A323997-1G A323997-250MG A323997-5G
76788-084EA 45.62 USD
76788-084 76788-086 77668-442
4-Phenylethynylbenzaldehyde 97%
4-Phenylethynylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₀O
MW: 206.25 g/mol
MDL Number: MFCD00114585
CAS Number: 57341-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR