Tri-(p-tert-butylphenyl)phosphate 95%

Supplier: Ambeed
Warning

A322926-1G A322926-250MG A322926-5G
76788-074EA 64 USD
76788-074 76788-076 76788-078
Tri-(p-tert-butylphenyl)phosphate 95%
Tri-(p-tert-butylphenyl)phosphate

Precaución: for Research Use Only

Formula: C₃₀H₃₉O₄P
MDL Number: MFCD00026302
CAS Number: 78-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR