(1S,2S,3R,4S,5S)-5-Amino-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetraol 97%

Supplier: Ambeed
Warning

Synonyms: Valiolamine

A319538-100MG A319538-1G A319538-250MG A319538-5G
76788-042EA 38 USD
76788-042 76788-044 77601-440 77668-416
(1S,2S,3R,4S,5S)-5-Amino-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetraol 97%
(1S,2S,3R,4S,5S)-5-Amino-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetraol

Precaución: for Research Use Only

Formula: C₇H₁₅NO₅
MDL Number: MFCD07773061
CAS Number: 83465-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR