3'-(Trifluoromethyl)biphenyl-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

A319375-1G A319375-250MG A319375-25G A319375-5G
76788-034EA 23.34 USD
76788-034 76788-036 76788-038 76788-040
3'-(Trifluoromethyl)biphenyl-4-carboxylic acid 98%
3'-(Trifluoromethyl)biphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O₂
MW: 266.22 g/mol
MDL Number: MFCD03424607
CAS Number: 195457-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR