6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A318162-100MG A318162-1G A318162-250MG A318162-5G
76788-018EA 12.74 USD
76788-018 76788-020 76788-022 76788-024
6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid 95%
6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MW: 242.0300 g/mol
MDL Number: MFCD09414724
CAS Number: 1000018-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR