(R)-N-Boc-3-methylmorpholine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-N-Boc-3-Methylmorpholine

A318081-100MG A318081-250MG
76788-014EA 202 USD
76788-014 76788-016
(R)-N-Boc-3-methylmorpholine 97%
(R)-N-Boc-3-methylmorpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.26 g/mol
MDL Number: MFCD16038029
CAS Number: 1022093-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR