6-Chloroquinoline-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Carboxy-6-chloroquinoline

A316037-1G A316037-250MG
76787-992EA 152 USD
76787-992 77872-443
6-Chloroquinoline-3-carboxylic acid 98%
6-Chloroquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₂
MDL Number: MFCD09787826
CAS Number: 118791-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR