Tolbutamide 98%

Supplier: Ambeed
Warning

Synonyms: N-(Butylcarbamoyl)-4-methylbenzenesulphonamide

A315619-1G A315619-5G A315619-100G
76787-988EA 5.31 USD
76787-988 76787-990 77668-388
Tolbutamide 98%
Tolbutamide

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₃S
MW: 270.35
MDL Number: MFCD00027169
CAS Number: 64-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR