Tetrakis(acetonitrile)palladium(II) tetrafluoroborate 98+%

Supplier: Ambeed
Warning

A315232-100MG A315232-1G A315232-250MG A315232-5G
76787-982EA 16 USD
76787-982 76787-984 76787-986 77872-442
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate 98+%

Precaución: for Research Use Only

Formula: C₈H₁₂B₂F₈N₄Pd
MDL Number: MFCD00043297
CAS Number: 21797-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR