N-Benzyloxycarbonyl-D-norvaline 98%

Supplier: Ambeed
Warning

Synonyms: N-Cbz-D-norvaline, Z-D-Nva-OH

A313432-1G A313432-250MG A313432-5G
76787-962EA 13 USD
76787-962 76787-964 76787-966
N-Benzyloxycarbonyl-D-norvaline 98%
N-Benzyloxycarbonyl-D-norvaline

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄
MW: 251.282 g/mol
MDL Number: MFCD00136826
CAS Number: 42918-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR