tert-Butyldimethylsilanol ≥99%

Supplier: Ambeed
Danger

A312242-5G A312242-1G
76787-954EA 10 USD
76787-954 77668-378
tert-Butyldimethylsilanol ≥99%
tert-Butyldimethylsilanol

Precaución: for Research Use Only

Formula: C₆H₁₆OSi
MDL Number: MFCD00010673
CAS Number: 18173-64-3
UN: 1987
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR