4-Chloro-1-methyl-1H-pyrazole 95%

Supplier: Ambeed
Warning

A312036-10G A312036-5G A312036-1G
76787-952EA 293.87 USD
76787-952 77601-406 77668-376
4-Chloro-1-methyl-1H-pyrazole 95%
4-Chloro-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅ClN₂
MW: 116.5500 g/mol
MDL Number: MFCD08275463
CAS Number: 35852-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR