2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-ɑ-D-glucopyranosyl chloride 93%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucose, 2-Acetamido-2-deoxy-α-D-glucopyranosyl chloride 3,4,6-triacetate, 2-Acetamido-2-deoxy-ɑ-D-glucopyranosyl Chloride 3,4,6-Triacetate, α-Acetochloro-D-glucosamine, 1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-ɑ-D-glucose, (2R,3S,4R,5R,6R)-5-Acetamido-2-(acetoxymethyl)-6-chlorotetrahydro-2H-pyran-3,4-diyl diacetate, 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-glucopyranosyl chloride

A311869-100MG A311869-1G A311869-5G A311869-250MG A311869-25G
76787-948EA 9 USD
76787-948 76787-950 77568-630 77668-374 77872-434
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-ɑ-D-glucopyranosyl chloride 93%
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-alpha-D-glucopyranosyl chloride

Precaución: For research use only.

Formula: C₁₄H₂₀ClNO₈
MDL Number: MFCD00069776
CAS Number: 3068-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR