trans,trans-4'-(3-Butenyl)-4-(p-tolyl)bicyclohexyl 97%

Supplier: Ambeed
Warning

Synonyms: (trans,trans)-4-(But-3-en-1-yl)-4'-(p-tolyl)-1,1'-bi(cyclohexane), trans,trans-4'-(3-Butenyl)-4-(4-methylphenyl)bicyclohexyl

A311669-100MG A311669-1G A311669-250MG A311669-25G A311669-5G
76787-938EA 5 USD
76787-938 76787-940 76787-942 76787-944 76787-946
trans,trans-4'-(3-Butenyl)-4-(p-tolyl)bicyclohexyl 97%
trans,trans-4'-(3-Butenyl)-4-(p-tolyl)bicyclohexyl

Precaución: for Research Use Only

Formula: C₂₃H₃₄
MDL Number: MFCD13188633
CAS Number: 129738-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR