N-Cbz-D-glutamine 98%

Supplier: Ambeed
Warning

Synonyms: Cbz-D-Gln-OH, ((Benzyloxy)carbonyl)-D-glutamine, Z-D-Gln-OH

A311188-100MG A311188-1G A311188-250MG A311188-25G A311188-5G
76787-910EA 5.31 USD
76787-910 76787-912 76787-914 76787-916 76787-918
N-Cbz-D-glutamine 98%
N-Cbz-D-glutamine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₅
MW: 280.28 g/mol
MDL Number: MFCD00065698
CAS Number: 13139-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR