trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl 97%

Supplier: Ambeed
Warning

Synonyms: (trans,trans)-4-(3,4-Difluorophenyl)-4'-vinyl-1,1'-bi(cyclohexane)

A310503-1G A310503-250MG A310503-25G A310503-5G
76787-898EA 11 USD
76787-898 76787-900 76787-902 76787-904
trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl 97%
trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl

Precaución: for Research Use Only

Formula: C₂₀H₂₆F₂
MW: 304.425 g/mol
MDL Number: MFCD09842838
CAS Number: 142400-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR