Sulphacetamide 98%

Supplier: Ambeed
Warning

Synonyms: N-((4-Aminophenyl)sulphonyl)acetamide

A308801-100MG A308801-500G A308801-1MG
76787-890EA 6 USD
76787-890 77601-396 77668-366
Sulphacetamide 98%
Sulphacetamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃S
MW: 214.24 g/mol
MDL Number: MFCD00066501
CAS Number: 144-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR