Sucralfate 30-38%

Supplier: Ambeed
Warning

Synonyms: Hexadeca-μ-hydroxytetracosahydroxy[μ8-[[1,3,4,6-tetra-O-sulpho-β-D-fructofuranosyl α-D-glucopyranoside tetrakis(sulphato-κO′)](8-)]]hexadecaaluminum

A308396-100G A308396-1G A308396-500G
76787-882EA 15.92 USD
76787-882 77601-388 77668-364
Sucralfate 30-38%
Sucralfate

Precaución: for Research Use Only

Formula: C₁₂H₅₄Al₁₆O₇₅S₈
MW: 2086.7368 g/mol
MDL Number: MFCD00145339
CAS Number: 54182-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR