PTC-124 98%

Supplier: Ambeed
Warning

A306827-100MG A306827-10G A306827-10MG A306827-1G A306827-250MG A306827-50MG A306827-5G A306827-5MG
76787-860EA 11 USD
76787-860 76787-862 76787-864 76787-866 76787-868 76787-870 76787-872 76787-874
PTC-124 98%
PTC-124

Precaución: for Research Use Only

Formula: C15H9FN2O3
MDL Number: MFCD09864996
CAS Number: 775304-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR