N-Benzyloxycarbonyl-D-phenylalaninamide 97%

Supplier: Ambeed
Warning

Synonyms: (R)-Benzyl (1-amino-1-oxo-3-phenylpropan-2-yl)carbamate

A306364-10G A306364-1G A306364-250MG
76787-850EA 52 USD
76787-850 76787-852 76787-854
N-Benzyloxycarbonyl-D-phenylalaninamide 97%
N-Benzyloxycarbonyl-D-phenylalaninamide

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₃
MW: 298.342 g/mol
MDL Number: MFCD00191136
CAS Number: 5241-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR