1-Boc-3-fluoro-4-piperidone 97%

Supplier: Ambeed
Warning

A304821-100G A304821-10G A304821-1G A304821-250MG A304821-25G A304821-5G
76787-816EA 198 USD
76787-816 76787-818 76787-820 76787-822 76787-824 76787-826
1-Boc-3-fluoro-4-piperidone 97%
1-Boc-3-fluoro-4-piperidone

Precaución: for Research Use Only

Formula: C₁₀H₁₆FNO₃
MDL Number: MFCD10566554
CAS Number: 211108-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR