2,3,5-Trimethylhydroquinone 98%

Supplier: Ambeed
Danger

Synonyms: 2,3,5-Trimethylbenzene-1,4-diol, Trimethylhydroquinone

A303606-25G A303606-1000G A303606-100G A303606-500G
76787-804EA 8.49 USD
76787-804 77601-368 77601-370 77668-346
2,3,5-Trimethylhydroquinone 98%
2,3,5-Trimethylhydroquinone

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00002346
CAS Number: 700-13-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR