(1-Pentyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: Pentyltriphenylphosphonium bromide

A303327-100G A303327-25G A303327-500G A303327-5G
76787-792EA 57 USD
76787-792 76787-794 76787-796 76787-798
(1-Pentyl)triphenylphosphonium bromide 97%
(1-Pentyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: H₃C(CH₂)₄P(C₆H₅)₃Br
MDL Number: MFCD00031630
CAS Number: 21406-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR