Undecanal 97%

Supplier: Ambeed
Warning

Synonyms: Hendecanal, Undecylaldehyde

A302937-100G A302937-25G A302937-500G
76787-780EA 31.83 USD
76787-780 76787-782 76787-784
Undecanal 97%
Undecanal

Precaución: for Research Use Only

Formula: C₁₁H₂₂O
MW: 170.3 g/mol
MDL Number: MFCD00007016
CAS Number: 112-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR