2-Chloro-3-fluoro-5-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A302101-100MG
A302101-10G
A302101-1G
A302101-250MG
A302101-5G
A302101-25G
76787-770EA
6
USD
76787-770
76787-772
76787-774
76787-776
76787-778
77668-338
2-Chloro-3-fluoro-5-nitropyridine 98%
2-Chloro-3-fluoro-5-nitropyridine
Precaución: for Research Use Only
Formula:
C₅H₂ClFN₂O₂ |
MDL Number:
MFCD16659664 CAS Number: 1079179-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...