Thiophene-2-carbonyl chloride 95%

Supplier: Ambeed
Danger

A301688-1G A301688-25G A301688-5G
76787-756EA 9 USD
76787-756 76787-758 76787-760
Thiophene-2-carbonyl chloride 95%
Thiophene-2-carbonyl chloride

Precaución: for Research Use Only

Formula: C₅H₃ClOS
MDL Number: MFCD00005428
CAS Number: 5271-67-0
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR