Methyl 2-(2-pyrimidyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(2-Pyrimidyl)acetate

A300780-1G A300780-250MG A300780-25G A300780-5G
76787-742EA 51 USD
76787-742 76787-744 76787-746 76787-748
Methyl 2-(2-pyrimidyl)acetate 98%
Methyl 2-(2-pyrimidyl)acetate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD12198887
CAS Number: 60561-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR