tert-Butyl 2,6-Diazaspiro[3.3]heptane-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 2-Boc-2,6-diazaspiro[3.3]heptane, tert-Butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate, 2-(tert-Butoxycarbonyl)-2,6-diazaspiro[3.3]heptane, 2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-Butyl ester

A300058-100MG A300058-1G A300058-250MG A300058-5G
76787-736EA 59.42 USD
76787-736 76787-738 76787-740 77550-824
tert-Butyl 2,6-Diazaspiro[3.3]heptane-2-carboxylate 95%
tert-Butyl 2,6-Diazaspiro[3.3]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
MW: 198.27 g/mol
MDL Number: MFCD11226963
CAS Number: 1041026-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR