AG-494 95%

Supplier: Ambeed
Danger

Synonyms: Tyrphostin AG 494, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-phenylacrylamide

A299892-100MG A299892-10MG A299892-1MG A299892-50MG A299892-5MG
76787-726EA 87 USD
76787-726 76787-728 76787-730 76787-732 76787-734
AG-494 95%
AG-494

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂O₃
MW: 280.3 g/mol
MDL Number: MFCD00209863
CAS Number: 133550-35-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR