5-Bromo-4-chloro-2-(methylthio)pyrimidine 98%

Supplier: Ambeed
Danger

Synonyms: 5-bromo-4-chloro-2-(methylsulphanyl)pyrimidine

A299804-10G A299804-1G A299804-25G A299804-5G
76787-718EA 28 USD
76787-718 76787-720 76787-722 76787-724
5-Bromo-4-chloro-2-(methylthio)pyrimidine 98%
5-Bromo-4-chloro-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂S
MW: 239.5200 g/mol
MDL Number: MFCD00023232
CAS Number: 63810-78-6
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR