5-Bromo-2-phenylpyrimidine 95%

Supplier: Ambeed
Warning

A299474-10G A299474-1G A299474-250MG A299474-5G
76787-698EA 267 USD
76787-698 76787-700 76787-702 76787-704
5-Bromo-2-phenylpyrimidine 95%
5-Bromo-2-phenylpyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₂
MDL Number: MFCD09999220
CAS Number: 38696-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR