2-Fluoro-3-nitrophenol 97%

Supplier: Ambeed
Danger

A298741-10G A298741-1G A298741-5G A298741-100MG
76787-678EA 310 USD
76787-678 76787-680 76787-682 77872-393
2-Fluoro-3-nitrophenol 97%
2-Fluoro-3-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄FNO₃
MDL Number: MFCD11042443
CAS Number: 179816-26-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR