2'-(Trifluoromethyl)biphenyl-4-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

A298667-1G A298667-250MG A298667-5G
76787-672EA 81.69 USD
76787-672 76787-674 76787-676
2'-(Trifluoromethyl)biphenyl-4-carboxylic acid 95%
2'-(Trifluoromethyl)biphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O₂
MW: 266.22 g/mol
MDL Number: MFCD03424605
CAS Number: 198205-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR