2,5-Dioxopyrrolidin-1-yl stearate 95%

Supplier: Ambeed
Warning

A295188-1G A295188-5G A295188-250MG
76787-622EA 23 USD
76787-622 77668-314 77872-384
2,5-Dioxopyrrolidin-1-yl stearate 95%
2,5-Dioxopyrrolidin-1-yl stearate

Precaución: for Research Use Only

Formula: C₂₂H₃₉NO₄
MDL Number: MFCD00050403
CAS Number: 14464-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR