(R)-Benzyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate 96%

Supplier: Ambeed
Warning

A294479-5G A294479-1G
76787-620EA 149.59 USD
76787-620 77668-312
(R)-Benzyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate 96%
(R)-Benzyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MW: 235.28 g/mol
MDL Number: MFCD02683223
CAS Number: 72597-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR