6-Fluorospiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one 97%

Supplier: Ambeed
Warning

A294411-100MG A294411-250MG
76787-616EA 26.53 USD
76787-616 76787-618
6-Fluorospiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one 97%
6-Fluorospiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₃FN₂O₂
MW: 236.2400 g/mol
MDL Number: MFCD10566063
CAS Number: 92926-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR