Trospium chloride 99%

Supplier: Ambeed
Warning

Synonyms: rel-(1R,3r,5S)-3-(2-Hydroxy-2,2-diphenylacetoxy)spiro[bicyclo[3.2.1]octane-8,1'-pyrrolidin]-1'-ium chloride

A293957-1G A293957-1MG A293957-250MG A293957-50MG A293957-100MG
76787-604EA 81.69 USD
76787-604 76787-606 76787-608 76787-610 77668-308
Trospium chloride 99%
Trospium chloride

Precaución: for Research Use Only

Formula: C₂₅H₃₀ClNO₃
MW: 427.97 g/mol
MDL Number: MFCD00865254
CAS Number: 10405-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR