Spiro[indene-1,4'-piperidine] 98%

Supplier: Ambeed
Warning

A293403-100MG A293403-1G A293403-250MG A293403-5G
76787-588EA 28.65 USD
76787-588 76787-590 76787-592 77568-602
Spiro[indene-1,4'-piperidine] 98%
Spiro[indene-1,4'-piperidine]

Precaución: For research use only.

Formula: C₁₃H₁₅N
MW: 185.26 g/mol
MDL Number: MFCD08705802
CAS Number: 33042-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR