Trimethylamine-N-oxide 98%

Supplier: Ambeed
Warning

Synonyms: Trimethylamine N-oxide, Trimethylamine oxide, TMANO

A292360-1G A292360-250MG A292360-25G A292360-5G
76787-574EA 7 USD
76787-574 76787-576 76787-578 76787-580
Trimethylamine-N-oxide 98%
Trimethylamine N-oxide

Precaución: for Research Use Only

Formula: C₃H₉NO
MDL Number: MFCD00002048
CAS Number: 1184-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR