Ethyl-6-amino-2-phenyl-1H-indole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-amino-2-phenyl-1H-indole-3-carboxylate

A292273-100MG A292273-10G A292273-1G
76787-568EA 52 USD
76787-568 76787-570 76787-572
Ethyl-6-amino-2-phenyl-1H-indole-3-carboxylate 95%
Ethyl-6-amino-2-phenyl-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂O₂
MDL Number: MFCD09953516
CAS Number: 945655-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR