Vitamin A acetate 2800000 IU/g

Supplier: Ambeed
Danger

Synonyms: all-trans-Retinyl acetate, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate

A290342-1G A290342-25G A290342-5G A290342-10G A290342-500G
76787-540EA 5.31 USD
76787-540 76787-542 76787-544 77601-292 77668-286
Vitamin A acetate 2800000 IU/g
all-trans-Retinyl acetate

Precaución: for Research Use Only

Formula: C₂₂H₃₂O₂
MW: 328.49 g/mol
MDL Number: MFCD00019413
CAS Number: 127-47-9
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR