Tetraethylammonium trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Tetraethylammoniumtrifluoromethanesulphonate

A288822-100G A288822-1G A288822-25G A288822-5G
76787-504EA 277.96 USD
76787-504 76787-506 76787-508 77593-772
Tetraethylammonium trifluoromethanesulfonate 98%
Tetraethylammonium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₉H₂₀F₃NO₃S
MW: 279.3202 g/mol
MDL Number: MFCD00042586
CAS Number: 35895-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR