2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Phosphoric Acid 2-(Methacryloyloxy)ethyl 2-(Trimethylammonio)ethyl Ester, 2-(Methacryloyloxy)ethyl (2-(trimethylammonio)ethyl) phosphate, 2-Methacryloyloxyethyl Phosphorylcholine
A287632-25G
A287632-5G
A287632-100G
A287632-1G
76787-482EA
288
USD
76787-482
76787-484
77601-274
77601-276
2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate 98%
2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate
Precaución: for Research Use Only
Formula:
C₁₁H₂₂NO₆P |
MDL Number:
MFCD11112180 CAS Number: 67881-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...