(S)-2-(((Benzyloxy)carbonyl)amino)butanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Z-Abu-OH, Cbz-Abu-OH, (S)-2-(Benzyloxycarbonylamino)butyric acid

A286860-1G A286860-25G A286860-5G
76787-472EA 5.31 USD
76787-472 76787-474 76787-476
(S)-2-(((Benzyloxy)carbonyl)amino)butanoic acid 95%
(S)-2-(((Benzyloxy)carbonyl)amino)butanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MW: 237.3 g/mol
MDL Number: MFCD00077006
CAS Number: 42918-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR